ENAH11DD1535 Enamine

6,6-difluoro-5,6,7,8,9,10-hexahydrobenzo[8]annulen-5-one

  • MDL number MFCD31665693
  • Cas Number : 1238627-55-8
  • Molecular Weight : 210.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1706524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1706524
Availability details for
Quantity
  • Availability
  • Add to Cart