ENAH11DD156D Enamine

3-chloro-4,4,4-trifluorobutanoic acid

  • MDL number MFCD31666024
  • Cas Number : 2228824-02-8
  • Molecular Weight : 176.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1709876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1709876
Availability details for
Quantity
  • Availability
  • Add to Cart