ENAH11DD1577 Enamine

tert-butyl N-[(1-amino-2,3-dihydro-1H-inden-5-yl)methyl]carbamate hydrochloride

  • MDL number MFCD31666138
  • Cas Number : 2225154-18-5
  • Molecular Weight : 298.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1710074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1710074
Availability details for
Quantity
  • Availability
  • Add to Cart