ENAH11DD15EA Enamine

ethyl 6,6-dimethyl-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carboxylate

  • MDL number MFCD31666020
  • Cas Number : 2107179-94-0
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1706047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1706047
Availability details for
Quantity
  • Availability
  • Add to Cart