ENAH11DD16E5 Enamine

1-(quinolin-5-yl)ethan-1-one

  • MDL number MFCD09754106
  • Cas Number : 103854-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1709729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1709729
Availability details for
Quantity
  • Availability
  • Add to Cart