ENAH11DD16FC Enamine

(3-fluoro-4-methylpyridin-2-yl)methanamine dihydrochloride

  • MDL number MFCD31666042
  • Cas Number : 312905-00-3
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1709833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1709833
Availability details for
Quantity
  • Availability
  • Add to Cart