ENAH11DD1787 Enamine

3-[(methylamino)methyl]-4,5-dihydro-1H-1,2,4-triazol-5-one hydrochloride

  • MDL number MFCD06738809
  • Cas Number : 1187928-77-3
  • Molecular Weight : 164.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1716006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1716006
Availability details for
Quantity
  • Availability
  • Add to Cart