ENAH11DD17A2 Enamine

2-[(tert-butoxy)carbonyl]-2-azaspiro[4.5]decane-8-carboxylic acid

  • MDL number MFCD22566121
  • Cas Number : 1363381-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1716084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1716084
Availability details for
Quantity
  • Availability
  • Add to Cart