ENAH11DD17AA Enamine

(2E)-2-fluoro-3-(1H-imidazol-5-yl)prop-2-enoic acid hydrochloride

  • MDL number MFCD31665896
  • Molecular Weight : 192.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1716121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1716121
Availability details for
Quantity
  • Availability
  • Add to Cart