ENAH11DD17FB Enamine

6-oxo-1-[4-(propan-2-yl)phenyl]-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD31666386
  • Cas Number : 2225144-20-5
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1716730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1716730
Availability details for
Quantity
  • Availability
  • Add to Cart