ENAH11DD1995 Enamine

1-(cyclopropylmethyl)azetidin-3-amine; bis(trifluoroacetic acid)

  • MDL number MFCD31665497
  • Cas Number : 2225141-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718223
Availability details for
Quantity
  • Availability
  • Add to Cart