ENAH11DD199E Enamine

8-bromo-1,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD31690109
  • Cas Number : 1260221-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718395
Availability details for
Quantity
  • Availability
  • Add to Cart