ENAH11DD19A7 Enamine

1-(3-fluoropyridin-4-yl)-3-methylpiperazine dihydrochloride

  • MDL number MFCD31666264
  • Cas Number : 2225142-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718541
Availability details for
Quantity
  • Availability
  • Add to Cart