ENAH11DD19B0 Enamine

[4-iodo-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methanamine hydrochloride

  • MDL number MFCD31665659
  • Cas Number : 2225141-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718727
Availability details for
Quantity
  • Availability
  • Add to Cart