ENAH11DD19C0 Enamine

(2R)-1-(3-chloro-2,6-difluorophenyl)propan-2-ol

  • MDL number MFCD31666031
  • Cas Number : 2227679-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718936
Availability details for
Quantity
  • Availability
  • Add to Cart