ENAH11DD1A52 Enamine

3-fluoro-1-methylcyclobutan-1-amine hydrochloride

  • MDL number MFCD31618008
  • Cas Number : 1781122-54-0
  • Molecular Weight : 139.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1718242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718242
Availability details for
Quantity
  • Availability
  • Add to Cart