ENAH11DD1A64 Enamine

methyl 4-oxo-4H,5H-thieno[3,2-c]quinoline-2-carboxylate

  • MDL number MFCD31665177
  • Cas Number : 1469294-49-2
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1717055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717055
Availability details for
Quantity
  • Availability
  • Add to Cart