ENAH11DD1A70 Enamine

tert-butyl 3-(aminomethyl)-4-methylpiperidine-1-carboxylate

  • MDL number MFCD24531569
  • Cas Number : 1520880-67-4
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1718387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718387
Availability details for
Quantity
  • Availability
  • Add to Cart