ENAH11DD1A78 Enamine

[(3aR,6aS)-6a-(hydroxymethyl)-octahydrocyclopenta[c]pyrrol-3a-yl]methanol hydrochloride

  • MDL number MFCD31665577
  • Cas Number : 2225127-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718428
Availability details for
Quantity
  • Availability
  • Add to Cart