ENAH11DD1ACB Enamine

ethyl (2S)-2-amino-3-cyclohexylpropanoate hydrochloride

  • MDL number MFCD31666272
  • Cas Number : 60025-25-4
  • Molecular Weight : 235.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1718730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718730
Availability details for
Quantity
  • Availability
  • Add to Cart