ENAH11DD1B67 Enamine

ethyl 5-(cyanomethyl)-1-methyl-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD31666086
  • Cas Number : 2225137-23-3
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1717115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717115
Availability details for
Quantity
  • Availability
  • Add to Cart