ENAH11DD1BB2 Enamine

tert-butyl 3-(5-amino-1-methyl-1H-1,2,4-triazol-3-yl)azetidine-1-carboxylate

  • MDL number MFCD31666794
  • Cas Number : 2228572-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1719474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1719474
Availability details for
Quantity
  • Availability
  • Add to Cart