ENAH11DD1BB9 Enamine

N-{14-iodo-1,10-diazatricyclo[9.4.0.0,3,8]pentadeca-3(8),4,6,10,12,14-hexaen-9-ylidene}prop-2-enamide

  • Cas Number : 326619-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1719499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1719499
Availability details for
Quantity
  • Availability
  • Add to Cart