ENAH11DD1BFC Enamine

3-(2,5-dichlorobenzenesulfonyl)propanoic acid

  • MDL number MFCD00450486
  • Cas Number : 864233-50-1
  • Molecular Weight : 283.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1719757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1719757
Availability details for
Quantity
  • Availability
  • Add to Cart