ENAH11DD1C4A Enamine

3-(phenoxymethyl)-4H-1,2,4-triazole

  • MDL number MFCD31665176
  • Cas Number : 856861-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717044
Availability details for
Quantity
  • Availability
  • Add to Cart