ENAH11DD1C4F Enamine

2-amino-N-(2,6-dimethylphenyl)-N-(propan-2-yl)acetamide

  • MDL number MFCD31665251
  • Cas Number : 71359-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717157
Availability details for
Quantity
  • Availability
  • Add to Cart