ENAH11DD1C53 Enamine

6-chloro-3-iodo-5-nitro-1H-indazole

  • MDL number MFCD08690112
  • Cas Number : 1000342-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717767
Availability details for
Quantity
  • Availability
  • Add to Cart