ENAH11DD1C55 Enamine

(3aR,6aS)-hexahydro-[1,3]dioxolo[4,5-c]pyrrole hydrochloride

  • MDL number MFCD19690585
  • Cas Number : 692058-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717795
Availability details for
Quantity
  • Availability
  • Add to Cart