ENAH11DD1C6E Enamine

ethyl 2-amino-1-methylcyclohexane-1-carboxylate hydrochloride

  • MDL number MFCD31666452
  • Cas Number : 2225136-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720104
Availability details for
Quantity
  • Availability
  • Add to Cart