ENAH11DD1C85 Enamine

(1R)-1-(4-bromo-3-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD24417187
  • Cas Number : 2225126-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720197
Availability details for
Quantity
  • Availability
  • Add to Cart