ENAH11DD1C92 Enamine

4-({[4-ethyl-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}methyl)-3-nitrobenzamide

  • MDL number MFCD11772834
  • Cas Number : 1110949-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720271
Availability details for
Quantity
  • Availability
  • Add to Cart