ENAH11DD1C98 Enamine

tert-butyl N-(2,2-difluoroethyl)-N-(piperidin-3-yl)carbamate

  • MDL number MFCD28537308
  • Cas Number : 1783709-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718146
Availability details for
Quantity
  • Availability
  • Add to Cart