ENAH11DD1C9A Enamine

4-[4-({[(tert-butoxy)carbonyl]amino}methyl)phenyl]benzoic acid

  • MDL number MFCD29917512
  • Cas Number : 193152-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720325
Availability details for
Quantity
  • Availability
  • Add to Cart