ENAH11DD1C9B Enamine

4-methyl-4-(4-methylphenyl)piperidine

  • MDL number MFCD18277107
  • Cas Number : 1268093-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718149
Availability details for
Quantity
  • Availability
  • Add to Cart