ENAH11DD1C9D Enamine

3-{4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]phenyl}propanoic acid

  • MDL number MFCD31666716
  • Cas Number : 919528-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720341
Availability details for
Quantity
  • Availability
  • Add to Cart