ENAH11DD1CA2 Enamine

1-[3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)phenyl]cyclobutane-1-carboxylic acid

  • MDL number MFCD31666852
  • Cas Number : 2229448-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720377
Availability details for
Quantity
  • Availability
  • Add to Cart