ENAH11DD1CA3 Enamine

4-{2-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]phenyl}benzoic acid

  • MDL number MFCD31666780
  • Cas Number : 215248-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720385
Availability details for
Quantity
  • Availability
  • Add to Cart