ENAH11DD1CA5 Enamine

tert-butyl(chloromethoxy)dimethylsilane

  • MDL number MFCD20621945
  • Cas Number : 119451-80-8
  • Molecular Weight : 180.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1718158

Safety Information

signalword Danger
pcodes P201,P202,P210,P233,P240,P241,P242,P243,P260,P264,P280,P281,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P308+P313,P310,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1718158
pictograms GHS02,GHS05,GHS08
hstatements H226,H314,H351
Availability details for
Quantity
  • Availability
  • Add to Cart