ENAH11DD1CF7 Enamine

2-{[(tert-butoxy)carbonyl]amino}-3-(1H-pyrrol-3-yl)propanoic acid

  • MDL number MFCD22217605
  • Cas Number : 1522017-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718199
Availability details for
Quantity
  • Availability
  • Add to Cart