ENAH11DD1CFA Enamine

5-(chlorosulfonyl)-2,4-dimethylfuran-3-carboxylic acid

  • MDL number MFCD31665491
  • Cas Number : 1507321-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1718201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1718201
Availability details for
Quantity
  • Availability
  • Add to Cart