ENAH11DD1CFE Enamine

tert-butyl 2-amino-3-cyclohexylpropanoate hydrochloride

  • MDL number MFCD18252695
  • Cas Number : 135837-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720610
Availability details for
Quantity
  • Availability
  • Add to Cart