ENAH11DD1D02 Enamine

1-(4-tert-butylphenyl)-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD31665253
  • Cas Number : 2225136-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717174
Availability details for
Quantity
  • Availability
  • Add to Cart