ENAH11DD1D22 Enamine

(3-chloro-2-methylphenyl)methanamine hydrochloride

  • MDL number MFCD11045185
  • Cas Number : 202522-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720709
Availability details for
Quantity
  • Availability
  • Add to Cart