ENAH11DD1E3C Enamine

tert-butyl N-{[1-(aminomethyl)spiro[2.3]hexan-1-yl]methyl}carbamate

  • MDL number MFCD31690725
  • Cas Number : 2228153-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720763
Availability details for
Quantity
  • Availability
  • Add to Cart