ENAH11DD1E55 Enamine

(1Z)-buta-1,3-diene-1-sulfonamide

  • MDL number MFCD31690878
  • Cas Number : 83170-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721032
Availability details for
Quantity
  • Availability
  • Add to Cart