ENAH11DD1E59 Enamine

4-(aminomethyl)-2-methylphenol hydrobromide

  • MDL number MFCD31665137
  • Cas Number : 2225136-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721122
Availability details for
Quantity
  • Availability
  • Add to Cart