ENAH11DD1ECD Enamine

1-(2-chlorophenyl)-4-(3-chloropropyl)piperazine dihydrochloride

  • MDL number MFCD31666297
  • Cas Number : 2228135-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722413
Availability details for
Quantity
  • Availability
  • Add to Cart