ENAH11DD1EE4 Enamine

2-[3-(tert-butoxy)spiro[3.3]heptan-1-yl]acetic acid

  • MDL number MFCD31666379
  • Cas Number : 2228811-43-4
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722686
Availability details for
Quantity
  • Availability
  • Add to Cart