ENAH11DD1F45 Enamine

2-[(3R)-pyrrolidin-3-yloxy]acetic acid hydrochloride

  • MDL number MFCD31666879
  • Cas Number : 2227692-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724124
Availability details for
Quantity
  • Availability
  • Add to Cart