ENAH11DD1F50 Enamine

3-(2-fluorophenyl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD28108569
  • Cas Number : 2225136-99-0
  • Molecular Weight : 217.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1720916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720916
Availability details for
Quantity
  • Availability
  • Add to Cart